missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DraI (10 U/μL)

Product Code. 10679880 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1,500 units
Unit Size:
1500 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10679880 1,500 units 1500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10679880 Supplier Thermo Scientific™ Supplier No. ER0221
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific™ DraI is available as a FastDigest™ enzyme which is used for rapid DNA digestion.

Thermo Scientific™ DraI is available as a FastDigest™ enzyme which is used for rapid DNA digestion.

5'...T T T▵A A A...3'

3'...A A A▵T T T...5'

Conditions for 100% Activity:

  • 1X Buffer Tango™:
  • 33mM Tris-acetate (pH7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • DraI is supplied in:10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton X-100, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with DraI, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: never overlaps — no effect
  • EcoKI: may overlap — blocked
  • EcoBI: never overlaps — no effect

Digestion of Agarose-embedded DNA:

  • Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16hours
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme Dra I
Compatible Buffer 10X Buffer Tango
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 1,500 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.