missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Gel Extraction Kit

Product Code. 10670091 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 Preps
50 Preps
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10670091 250 Preps Each
10765651 50 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10670091 Supplier Thermo Scientific™ Supplier No. K0692

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific GeneJET Gel Extraction Kit is designed for rapid and efficient purification of DNA fragments from standard or low-melting point agarose gels run in either TAE or TBE buffer.

Thermo Scientific GeneJET Gel Extraction Kit is designed for rapid and efficient purification of DNA fragments from standard or low-melting point agarose gels run in either TAE or TBE buffer.

The kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column. The kit can be used to purify DNA fragments from 25 bp to 20 kb in size with recovery rates up to 95%. Each GeneJET purification column has a binding capacity of up to 25 μg of DNA and can process up to 1 g of agarose gel. The entire procedure takes just 15 minutes, and the isolated DNA is ready to use in all common downstream applications, including ligation, restriction digestion, PCR, sequencing, and labeling.

Highlights

  • Highly efficient: up to 95% recoveries in the range of 100 bp to 10 kb
  • Fast: procedure takes only 15 minutes
  • Convenient: spin columns are capped and assembled with collection tubes

Applications

Fast and efficient extraction of high purity DNA fragments ideal for use in all conventional molecular biology procedures including:

  • FastDigest or conventional restriction digestion
  • Automated fluorescent or radioactive sequencing
  • PCR
  • In situ hybridization
  • Labeling
  • Cloning
TRUSTED_SUSTAINABILITY

Specifications

Column Type Spin Column
Content And Storage For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
Isolation Technology Silica Spin Column
Sample Type Agarose gels
Elution Volume 50 μL
Final Product Type DNA
For Use With (Application) PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput Compatibility Not High-throughput Compatible (Manual)
Quantity 250 Preps
Shipping Condition Room Temperature
Test Time 15 min.
Yield 25 μg (Binding capacity)
Starting Material Amount ≤1 g
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • genejet gel extr 250 prep
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: thiocyanic acid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.