missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plant Genomic DNA Purification Kit

Product Code. 11581645 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 Preps
50 Preps
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11581645 250 Preps Each
11571645 50 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11581645 Supplier Thermo Scientific™ Supplier No. K0792
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types.

GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types. It utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types. It utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

Highlights
Fast—procedure completed in 30 min following cell lysis
Convenient—protocol does not include clogging-prone lysate filtration step
High yield—up to 32 μg of high quality DNA from 100 mg sample, including nuclear, mitochondrial, and chloroplast DNA
Compatible—use with awide variety of samples (leaves, roots, sprouts and other parts of various plants)

The standard procedure takes less than 30 minutes following cell lysis and yields purified DNA greater than 30 kb in size. DNA yields vary between different species and tissues depending on genome size, ploidy, cell number, and the age of the tissue sample. The typical yield from a standard tissue source (e. g. young wheat leaves) is 30 to 32 μg from 100 mg of tissue. Isolated DNA can be used directly in PCR, qPCR, southern blotting, and enzymatic reactions.

Applications
• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting

TRUSTED_SUSTAINABILITY

Specifications

Column Type Spin Column
Content And Storage • 6 x 1 mL RNase A; store at room temperature until opened, –20°C after opening
• 120 mL Lysis Buffer A; room temperature
• 15 mL Lysis Buffer B; room temperature
• 40 mL Precipitation Solution; room temperature
• 120 mL Plant gDNA Binding Solution; room temperature
• 40 mL Wash Buffer I (concentrated); room temperature
• 40 mL Wash Buffer II (concentrated); room temperature
• 150 mL Elution Buffer; room temperature
• 250 GeneJET Purification Columns with Collection Tubes; room temperature
• 250 Collection Tubes; room temperature

Isolation Technology Silica Spin Column
Scale Mini
Sample Type Plants
Elution Volume 200 μL
Final Product Type gDNA
For Use With (Application) PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput Compatibility Not High-throughput Compatible (Manual)
Quantity 250 Preps
Shipping Condition Room Temperature
Test Time 30 min. (After sample homogenization)
Yield ≤32 μg per 100 mg tissue
Starting Material Amount ≤100 mg of fresh/frozen
≤20 mg lyophilized
Show More Show Less
compliance-icons
Product Identifier
  • GeneJET Plant Genomic DNA Purification Kit
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium lauryl sulfate, Guanidine hydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.