missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plasmid Maxiprep Kit

Product Code. p-4530571 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
10 Preps
25 Preps
Unit Size:
25 preps
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
11551645 10 Preps Each
11561645 25 Preps 25 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11551645 Supplier Thermo Scientific™ Supplier No. K0491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific GeneJET Plasmid Maxiprep Kit, sufficient for 25 preps, utilizes silica-based membrane technology in the form of convenient spin column.

Thermo Scientific GeneJET Plasmid Maxiprep Kit, sufficient for 25 preps, utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers greater than 400 ng/μL of high copy plasmid DNA that can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

Highlights

  • Efficient—high yields up to 750 μg of high quality plasmid DNA from 250 mL of bacterial culture
  • High concentration of DNA purified— >400 ng/μL
  • Convenient—spin column format, centrifuge or vacuum protocols
  • Fast—procedure takes 60 minutes, considerably faster than using gravity flow columns

Applications

Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:

  • Transfection of robust cell lines
  • FastDigest restriction enzyme digestion
  • Automated fluorescent sequencing
  • PCR
  • Cloning
  • Transformation
  • In vitro transcription
TRUSTED_SUSTAINABILITY

Specifications

Concentration 400 ng/μL
Column Type Spin Column
Format Spin Column
Isolation Technology Silica Column
Scale Maxi
Sample Type Bacteria
For Use With (Application) Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 10 Preps
Prep Scale > 200 μg (Large-Scale) Plasmid DNA
Product Line GeneJET
Product Type Plasmid MaxiPrep Kit
Quantity 10 preps
Target Plasmid DNA
Test Time 60 min.
Show More Show Less
compliance-icons
Product Identifier
  • GeneJET Plasmid Maxiprep Kit
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sodium lauryl sulphate, sodium hydroxide, acetic acid, guanidine hydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.