missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plasmid Midiprep Kit

Product Code. 11531645 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Preps
25 Preps
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11531645 25 Preps Each
11541645 100 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11531645 Supplier Thermo Scientific™ Supplier No. K0481

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Designed for large-scale isolation of high quality plasmid DNA from recombinant E.coli cultures.

Thermo Scientific GeneJET Plasmid Midiprep Kit, sufficient for 25 preps, is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures. The kit utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 200 μg of high copy number plasmid DNA. The purified plasmid DNA can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

Highlights

  • Efficient—high yields up to 200 μg of high quality plasmid DNA from 50 mL of bacterial culture
  • High concentration of DNA purified— >400 ng/μL
  • Convenient—spin column format, centrifuge or vacuum protocols
  • Fast—procedure takes 60 minutes, considerably faster than using gravity flow columns

Applications

Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:

  • FastDigest restriction enzyme digestion
  • Automated fluorescent sequencing
  • PCR
  • Cloning
  • Transformation
  • In vitro transcription
  • Transfection of robust cell lines
TRUSTED_SUSTAINABILITY

Specifications

Concentration 400 ng/μL
Column Type Spin Column
Format Spin Column
Isolation Technology Silica Column
Scale Midi
Sample Type Bacteria
For Use With (Application) Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR
High-throughput Compatibility Not High-throughput Compatible (Manual)
Prep Scale 100-200 μg (Medium-Scale) Plasmid DNA
Product Line GeneJET
Product Type Plasmid MidiPrep Kit
Quantity 25 Preps
Target Plasmid DNA
Test Time 60 min.
Show More Show Less
compliance-icons
Product Identifier
  • GeneJET Plasmid Midiprep Kit
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor / soap
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: guanidine hydrochloride, sodium lauryl sulphate, sodium hydroxide, acetic acid,triton X-100

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.