missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Whole Blood Genomic DNA Purification Mini Kit

Product Code. 11521655 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 Preps
50 Reactions
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11521655 50 Reactions Each
11531655 250 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11521655 Supplier Thermo Scientific™ Supplier No. K0781
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Thermo Scientific GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting, and enzymatic reactions.

Thermo Scientific GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting, and enzymatic reactions.

Highlights
Efficient—high yields of high molecular weight genomic DNA
Fast—20 minute procedure following cell lysis
Convenient—spin columns are capped and assembled with collection tubes
Pure—DNA pure from enzyme inhibitors, RNA and proteins. Ready to use for all demanding applications and validated in real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests
Compatible—suitable for wide variety of samples like fresh or frozen anticoagulated mammalian blood (treated with EDTA, Citrate or Heparine), aged blood, buffy coat, bone marrow, and body fluids

Applications
• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting Includes Proteinase K Solution Lysis Solution Wash Buffer WB I (concentrated) Wash Buffer II (concentrated) Elution Buffer GeneJET Purification Columns (pre-assembled with collection tubes) Collection tubes Detailed Protocol

TRUSTED_SUSTAINABILITY

Specifications

Column Type Spin Column
Format Spin Column
Isolation Technology Spin Column
Scale Mini
Sample Type Blood
For Use With (Application) Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Sequencing, PCR
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line GeneJET
Product Type Genomic DNA Purification Kit
Quantity 50 Reactions
Target Genomic DNA
Test Time <20 min.
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Are MagJET and GeneJET blood nucleic acid purification kits compatible with animal blood?

Yes, the kits are compatible with animal blood for DNA/RNA purification.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

compliance-icons compliance-icons
Product Identifier
  • GeneJET Whole Blood Genomic DNA Purification Mini Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Proteinase, Guanidine hydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.