missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneRuler 1 kb Plus DNA Ladder

Product Code. 11813993 Shop All Thermo Scientific Products
Change view
Click to view available options
Kit Contents:
6X DNA Loading Dye, 10 x 1mL
6X DNA Loading Dye, 2 x 1mL
Unit Size:
Pack of 25
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kit Contents Unit Size
11813993 6X DNA Loading Dye, 10 x 1mL Pack of 25
11581625 6X DNA Loading Dye, 2 x 1mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11813993 Supplier Thermo Scientific™ Supplier No. SM1332
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels.

GeneRuler 1 kb Plus DNA Ladder 75 to 20,000 bp is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels. The ladder is a mixture of chromatography-purified individual DNA fragments.

GeneRuler 1 kb Plus DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase.

The ladder is supplied with 6X DNA Loading Dye for sample DNA.

The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler 1kb Plus DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase. The ladder is supplied with 6X DNA Loading Dye for sample DNA.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Ready to Load No
Size Range 75 to 20000 bp
Gel Compatibility Agarose Gels
Kit Contents 6X DNA Loading Dye, 10 x 1mL
Quantity 25 x 50 μg
Product Line GeneRuler
Product Type DNA Ladder
Are the Thermo Scientific DNA ladders compatible with GelRed and GelGreen gel stains?

The GelRed or GelGreen nucleic acid dyes can be used with our DNA Ladders. Nucleic acid binding dyes can affect DNA migration during electrophoresis, therefore we highly recommend post-staining of gels. Please follow the gel staining protocol provided by the manufacturer of nucleic acid gel dyes.

Which Thermo Scientific DNA ladders are optimal for fast electrophoresis separation?

For fast sizing and quantification of double-stranded DNA, we recommend using FastRuler DNA Ladders. These ladders are easily resolved in a short separation distance after an 8-14 min run.

What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?

Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).

What are the end functional groups of Thermo Scientific DNA and RNA ladders?

Thermo Scientific DNA and RNA ladders have 5′-phosphate and 3′-hydroxyl groups.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.