missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneRuler Express DNA Ladder, ready-to-use

Product Code. 10590841 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10590841 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10590841 Supplier Thermo Scientific™ Supplier No. SM1553
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Get fast separation (in 5 to 15 minutes at 23  V/cm) of double-stranded DNA fragments in agarose gels. Premixed with Loading Dye for direct loading onto gel.

GeneRuler Express DNA Ladder, ready-to-use, is designed for fast separation (in 5 to 15 minutes at 23 V/cm) of double-stranded DNA fragments in agarose gels (see Figure 1 and Table 1). The band pattern of express ladders is not affected by the composition of electrophoresis buffer, voltage, or gel percentage. The ladder is a mixture of chromatography-purified individual DNA fragments.

GeneRuler Express DNA Ladder, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading onto agarose gels.

The ladder is supplied with 6X DNA Loading Dye for sample DNA.

Highlights

  • Ideal for both DNA sizing and approximate quantification 
  • Ideal for fast separation under a wide range of electrophoresis conditions 
  • Sharp bands 
  • Bright reference bands (given in red)
  • Can be directly loaded and are stable at room temperature for 6 months 
  • Supplied with loading dye for sample DNA 
TRUSTED_SUSTAINABILITY

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Ready to Load Yes
Size Range 100 to 5000 bp
Gel Compatibility Agarose Gels
Quantity 50 μg
Product Line GeneRuler
Product Type DNA Ladder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.