missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ InsTAclone™ PCR Cloning Kit

Product Code. 10723114 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
30 Reactions
Unit Size:
30 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10723114 30 Reactions 30 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10723114 Supplier Thermo Scientific™ Supplier No. K1214A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Perform direct one-step cloning of PCR products with 3'-dA overhangs generated by Taq DNA polymerase and other thermostable DNA polymerases which lack proofreading activity.

Thermo Scientific™ InsTA system for direct one-step cloning of PCR products with 3'-dA overhangs (1) generated by Taq DNA polymerase and other thermostable DNA polymerases which lack proofreading activity

The high quality ready-to-use TA cloning vector pTZ57R/T was prepared by linearizing the pTZ57R plasmid with Eco32I and tailing with single ddT. The 3'-ddT overhangs at both ends of the cloning site prevent recircularization of the vector during ligation, resulting in high cloning yields and low background. To increase the speed, convenience and efficiency of cloning experiment, the InsTAclone PCR Cloning Kit has been combined with the unique TransformAid Bacterial Transformation Kit—a set of solutions for preparation of chemically competent E. coli cells. According to the protocol, ligation and preparation of competent cells is performed in parallel. Therefore, it takes only one hour from the completion of PCR to cell plating. Our transformation protocol is often faster than the transformation of commercially available competent cells. The DNA insert can be readily excised from the versatile polylinker of pTZ57R/T, sequenced using standard M13/pUC primers or in vitro transcribed with T7 RNA polymerase.
  • Fast cloning—approximately one hour from PCR completion to cell plating
  • High efficiency—more than 90% of the recombinant clones contain the target DNA
  • One-step procedure—additional modifications of PCR fragment are not required
  • Compatibility—compatible with Taq, Tth, Tfl and other non-proofreading DNA polymerases, as well as with Long PCR Enzyme Mix
  • Convenience of pTZ57R/T cloning vector:

  • —ready-to-use: linearized with Eco32I and 3'-ddT tailed,

  • —MCS designed for easy mapping and manipulation of the cloned insert,

  • —blue/white screening,

  • —M13/pUC primer sites for sequencing or colony PCR screening,

  • —T7 promoter for in vitro transcription of the insert

Recommended for:

Sequencing of cloned insert; in vitro transcription of insert DNA; TA cloning.

Note:

E.coli strains are not included. Lyophilized bacterial strains GM2163 (†M0099) and JM107 (†M0109) can be provided free of charge upon request, with any purchase. Suitable for all common laboratory E.coli strains.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Sequencing of cloned insert; in vitro transcription of insert DNA; TA cloning.
No. of Reactions 30
Product Type InsTAclone PCR Cloning Kit
Quantity 30 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.