missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima Probe qPCR Master Mix (2X), with separate ROX vial

Product Code. 12201547 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
1,000 Reactions
200 Reactions
Unit Size:
1000 reactions
200 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
12201547 1,000 Reactions 1000 reactions
11873923 200 Reactions 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 12201547 Supplier Thermo Scientific™ Supplier No. K0262
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Optimize qPCR with probe chemistry using standard cycling protocols with these ready-to-use qPCR master mixes.

Maxima Probe qPCR Master Mixes are ready-to-use solutions optimized for quantitative real-time PCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima Probe qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates.

Maxima Probe qPCR Master Mixes are compatible with most real-time thermal cyclers.

The master mix includes Maxima Hot Start Taq DNA polymerase and dNTPs in an optimized PCR buffer. ROX Solution is provided in a separate vial for use with machines that require ROX as a passive reference dye. Maxima Hot Start Taq DNA polymerase in combination with the optimized buffer ensures PCR specificity and sensitivity.dUTP is included in the mix for optional carryover contamination control using uracil-DNA glycosylase (UDG). The use of Maxima Probe qPCR Master Mix in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates.
  • Specificity—Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity—detects low copy number targets
  • ROX Solution provided in a separate tube
  • Wide linear range—accurate quantification across 9 orders of magnitude
  • Universal—can be used on most real-time PCR instruments
  • Reproducibility and convenience—ready-to-use 2X master mix

Recommended for:

Real-time PCR and RT-qPCR using sequence specific probes on most real-time PCR machines.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains:
• 10 x 1.25 mL Maxima Probe qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer; sufficient for 1000 x 25 μL reactions.
• 250 μL ROX solution, 50 μM
• 10 x 1.25 mL Nuclease-Free Water

Store at -20°C.
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) BioRad CFX96™, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, MJ Chromo4, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, MJ Opticon
Product Line Maxima
Product Type qPCR Master Mix
Quantity 1,000 reactions
Sufficient For 1000 Reactions at 25 μL
For Use With (Application) Gene Expression
No. of Reactions 1,000 Reactions
Sample Type DNA, dsDNA
Volume 10 x 1.25 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.