missing translation for 'onlineSavingsMsg'
Learn More

Maxima Maxima SYBR Green/Fluorescein qPCR Master Mix (2X)

Code produit 11833923 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
No. of Reactions:
1,000 Reactions
200 reactions
4,000 Reactions
Conditionnement:
1000 reactions
200 reactions
4000 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit No. of Reactions Conditionnement
11833923 200 reactions 200 reactions
11826863 4,000 Reactions 4000 reactions
11843923 1,000 Reactions 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11833923 Fournisseur Maxima Code fournisseur K0241
Black Friday Starts Here
Use promo code 27984
Afficher toutes les promotions
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Ready-to-use solution optimized for qPCR and 2-step RT-qPCR.

The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most real-time thermal cyclers.

  • Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity - detects low copy number targets
  • Wide linear range - accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience - ready-to-use 2X master mix
TRUSTED_SUSTAINABILITY

Spezifikation

Concentration 2X
Content And Storage Contains:
• 2 x 1.25 mL Maxima SYBR Green/Fluorescein qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase, dNTPs (also dUTP) and SYBR Green I in an optimized PCR buffer with fluorescein passive reference dye; sufficient for 200 x 25 μL reactions.
• 2 x 1.25 mL Nuclease-Free Water

Store at -20°C.
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5
Product Line Maxima
Product Type Real Time PCR SYBR Master Mix
Quantity 200 reactions
Sufficient For 200 Reactions of 25 μL
For Use With (Application) Gene Expression
No. of Reactions 200 reactions
Sample Type DNA, dsDNA
Volume 2 x 1.25 mL
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.