missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima SYBR Green/ROX qPCR Master Mix (2X)

Product Code. 11893913 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Reactions:
1,000 Reactions
200 reactions
4,000 Reactions
Unit Size:
1000 reactions
200 reactions
4000 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
11893913 4,000 Reactions 4000 reactions
11873913 200 reactions 200 reactions
11883913 1,000 Reactions 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11893913 Supplier Thermo Scientific™ Supplier No. K0223
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready-to-use solution optimized for qPCR and 2-step RT-qPCR.

The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most real-time thermal cyclers.

  • Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity - detects low copy number targets
  • Wide linear range - accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience - ready-to-use 2X master mix
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains:
• 4 x 12.5 mL Maxima SYBR Green/ROX qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase, dNTPs (also dUTP) and SYBR Green I in an optimized PCR buffer with ROX passive reference dye; sufficient for 4000 x 25 μL reactions.
• 2 x 30 mL Nuclease-Free Water

Store at -20°C.
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
Product Line Maxima
Product Type Real Time PCR SYBR Master Mix
Quantity 4,000 reactions
Sufficient For 4000 Reactions of 25 μL
For Use With (Application) Gene Expression
No. of Reactions 4,000 Reactions
Sample Type DNA, dsDNA
Volume 4 x 12.5 mL
Show More Show Less
Is the Maxima SYBR Green/ROX qPCR Master Mix (2X) (Cat. No. K0223, K0221, K0222) compatible with the ViiA 7 Real-Time PCR System (Cat. No. 4453542, 4453539)

No, the ViiA 7 Real-Time PCR System (Cat. No. 4453542, 4453539) is not compatible with the Maxima SYBR Green/ROX qPCR Master Mix (2X) (Cat. No. K0223).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.