missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MboI (10 U/μL)

Product Code. 10795451 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1,500 units
300 units
Unit Size:
1500 units
300 units
This item is not returnable. View return policy

Product Code. 10795451

Brand: Thermo Scientific™ ER0812

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MboI restriction enzyme recognizes ^GATC sites and cuts best at 37°C in R buffer (Isoschizomers: BfuCI, BssMI, BstKTI, BstMBI, DpnII, Kzo9I, NdeII, Sau3AIm).

Lambda DNA (dam-) digested with MboI, 1.4% agarose, 116 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'..▵G A T C ...3'

3'.. C T A G▵...5'

Conditions for 100% Activity:

  • 1X Buffer R: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • MboI is supplied in:10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with MboI, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: completely overlaps —blocked
  • Dcm: never overlaps —no effect
  • CpG: may overlap —no effect
  • EcoKI: never overlaps —no effect
  • EcoBI: may overlap —blocked

Note:

Assayed using lambda DNA (dam) (#SD0021). MboI is blocked by overlappingdam methylation. To avoiddam methylation, use adam,dcm- strain such as GM2163 (#M0099). MboI, Bsp143I and DpnI all recognize the same sequence but have different methylation sensitivities and cleavage sites.

TRUSTED_SUSTAINABILITY

Specifikationer

Concentration 10 U/μL
Methylation Sensitivity Dam Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme Mbo I
Compatible Buffer 10X Buffer R
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 1,500 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.