missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ O'GeneRuler™ Ready-to-use 100bp Plus DNA Ladder

Product Code. 10756221 Shop All Thermo Scientific Products
Change view
Click to view available options
Size Range:
100 to 1000 bp
100 to 3000 bp
Unit Size:
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Range unitSize
10756221 100 to 3000 bp 50µg
10314250 100 to 1000 bp 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10756221 Supplier Thermo Scientific™ Supplier No. SM1153

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.

Thermo Scientific™ O'GeneRuler 100 bp Plus DNA Ladder, ready-to-use, is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.

O'GeneRuler 100 bp Plus DNA Ladder, ready-to-use, is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments. O'GeneRuler 100 bp Plus DNA Ladder, ready-to-use, is premixed with 6X Orange DNA Loading Dye for direct loading on gel. 6X Orange DNA Loading Dye contains xylene cyanol FF and orange G. In a 1% agarose gel orange G dye migrates at 50 bp. The ladder is supplied with 6X Orange DNA Loading Dye for sample DNA.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Can be directly loaded and are stable at room temperature for 6 months
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Concentration 6X Loading Dye
Size Range 100 to 3000 bp
Form Liquid
Kit Contents 6X Orange DNA Loading Dye
Quantity 100 Applications
Product Type DNA Molecular Weight Marker

This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount of the product in internal research conducted by the buyer. The buyer cannot use this product or materials made by the employment of this product for Commercial Purposes. Commercial Purposes means any activity for consideration and may include, but is not limited to: (1) use of the product in manufacturing; (2) use of the product to provide a service, information, or data; (3) use of the product for therapeutic, diagnostic or prophylactic purposes; or (4) resale of the product, whether or not the product is resold for use in research. Further information on purchasing licenses under the above patents may be obtained by contacting the Business Development Department, Invitrogen Corporation, 5791 Van Allen Way, Carlsbad, CA 92008.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.