Learn More
Thermo Scientific™ O'GeneRuler DNA Ladder, Ready-to-Use 50-1000 bp
Shop All Thermo Scientific ProductsDescription
O'GeneRuler 50 bp DNA Ladder, ready-to-use is recommended for sizing and approximate quantification of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.
O'GeneRuler 50 bp DNA Ladder, ready-to-use is premixed with 6X Orange DNA Loading Dye for direct loading on gel. 6X Orange DNA Loading Dye contains xylene cyanol FF and orange G. In a 1% agarose gel, orange G dye migrates at 50 bp. Therefore, O'GeneRuler 50 bp DNA Ladder is ideal when visualization of small DNA fragments is important.
The ladder is supplied with 6X Orange DNA Loading Dye for sample DNA.
Highlights
- Ideal for both DNA sizing and approximate quantification
- Sharp bands
- Bright reference bands (given in red)
- Can be directly loaded and are stable at room temperature for 6 months
- Supplied with loading dye for sample DNA
Specifications
Specifications
| Color | Orange |
| Concentration | 6X Loading Dye |
| Form | Liquid |
| Kit Contents | 6X Orange DNA Loading Dye, 1.00mL |
| Quantity | 100 Applications |
| Product Type | DNA Molecular Weight Marker |
This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount of the product in internal research conducted by the buyer. The buyer cannot use this product or materials made by the employment of this product for Commercial Purposes. Commercial Purposes means any activity for consideration and may include, but is not limited to: (1) use of the product in manufacturing; (2) use of the product to provide a service, information, or data; (3) use of the product for therapeutic, diagnostic or prophylactic purposes; or (4) resale of the product, whether or not the product is resold for use in research. Further information on purchasing licenses under the above patents may be obtained by contacting the Business Development Department, Invitrogen Corporation, 5791 Van Allen Way, Carlsbad, CA 92008.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.