missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RiboRuler Low Range RNA Ladder

Product Code. 11893993 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 x 20 μL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11893993 5 x 20 μL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11893993 Supplier Thermo Scientific™ Supplier No. SM1831
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Size and quantify RNA with Thermo Scientific™ RiboRuler Low Range RNA Ladder, a mixture of 7 chromatography purified single-stranded RNA transcripts (100 to 1000 bp long).

RiboRuler Low Range RNA Ladder 100-1000 bp is a mixture of 7 chromatography purified single-stranded RNA transcripts (100-1000 bp long), produced from specific templates that contain a fragment of the pTZ19R polylinker and Lambda phage fragments.

Ladder is available in 1 mM EDTA, pH 6.0.

RiboRuler Low Range RNA Ladder is composed of chromatography-purified transcripts what makes them free from NTPs and RNA degradation products. The concentration of each RNA transcript is determined spectrophotometrically (see Photo for precise quantities). To minimize the possibility of contamination during the use the ladder is aliquotted in smaller volumes.

RiboRuler Low Range RNA Ladder is optimal for end-labeling using T4 Polynucleotide Kinase and is ideal for Northern blots.

Highlights

  • Sharp bands of uniform intensity 
  • Easy-to-remember band sizes and quantities
  • Approximate RNA quantification by predetermined concentration of ladder bands
  • Supplied with 2X RNA Loading Dye 
  • Stable for 6 months at -20°C 
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 5 x 20 μL RiboRuler Low Range RNA Ladder (in 1 mM EDTA, pH 6.0); sufficient for 50 uses
• 1 mL RNA Loading Dye

Store at -70°C or at -20°C (up to 6 months).

Concentration 2X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Ethidium Bromide
Size Range 100 to 1000 bp
Quantity 5 x 20 μL
Product Line RiboRuler
Product Type DNA Ladder
compliance-icons
Product Identifier
  • RiboRuler Low Range RNA Ladder
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Formamide

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.