missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RiboRuler High Range RNA Ladder, ready-to-use

Product Code. 11883993 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 x 40 μL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11883993 5 x 40 μL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11883993 Supplier Thermo Scientific™ Supplier No. SM1823

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Perform RNA sizing and quantification on native or denaturing gels with this ready-to-use RNA Ladder composed of 8 chromatography purified single-stranded RNA transcripts.

RiboRuler High Range RNA Ladder 200 to 6000 bases, ready-to-use is a mixture of 8 chromatography purified single-stranded RNA transcripts (200 to 6000 bp long), produced from specific templates that contain a fragment of the pTZ19R polylinker and Lambda phage fragments. Ladder is premixed with loading dye.

RiboRuler High Range RNA Ladder, ready-to-use, is composed of chromatography-purified transcripts that makes them free from NTPs and RNA degradation products. The concentration of each RNA transcript is determined spectrophotometrically (see photo for precise quantities). To minimize the possibility of contamination during the use, the ladder is aliquotted in smaller volumes.

Highlights

  • Sharp bands of uniform intensity
  • Easy-to-remember band sizes and quantities
  • Predetermined concentration of ladder bands allows for approximate RNA quantification
  • Supplied with 2X RNA Loading Dye
  • Stable for 6 months at -20°C
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Ethidium Bromide
Size Range 200 to 6000 bp
Quantity 5 x 40 μL
Product Line RiboRuler
Product Type High Range DNA Ladder
compliance-icons
Product Identifier
  • RiboRuler High Range RNA Ladder, ready-to-use
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, formamide

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.