missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNase H (5 U/μL)

Product Code. 10334860 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 units
500 units
Unit Size:
100 units
500 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10334860 100 units 100 units
10519310 500 units 500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10334860 Supplier Thermo Scientific™ Supplier No. EN0201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ribonuclease H (RNase H) is an endoribonuclease that specifically degrades the RNA strand in RNA-DNA hybrids.

 Ribonuclease H (Rnase H) specifically degrades the RNA strand in RNA-DNA hybrids. It does not hydrolyze the phosphodiester bonds within single-stranded and double-stranded DNA and RNA.

  • Does not hydrolyze the phosphodiester bonds within single-stranded and double-stranded DNA and RNA
Quality Control:
  • The absence of endo-, exodeoxyribonucleases and ribonucleases confirmed by appropriate quality tests

Source:

  • E.coli MRE-600 cells

Molecular Weight:

  • 18.4kDa monomer

Definition of Activity Unit:

  • One unit of the enzyme catalyzes the formation of 1nmol of acid soluble products in 20 min. at 37°C
  • Enzyme activity is assayed in the following mixture: 20mM Tris-HCl (pH 7.8), 40mM KCl, 8mM MgCl2, 1mM DTT, 24μM [3H]-poly(A)·poly(dT), 0.03mg/mL BSA, 4% (v/v) glycerol
Storage Buffer:
  • The enzyme is supplied in: 25mM HEPES-KOH (pH 8.0), 50mM KCl, 0.1mM EDTA, 1mM DTT, 0.1mg/mL BSA and 50% (v/v) glycerol

10X Reaction Buffer:

  • 200mM Tris-HCl (pH 7.8), 400mM KCl, 80mM MgCl2, 10mM DTT

Inhibition and Inactivation:

  • Inhibitors: metal chelators, SH-blocking reagents
  • Inactivated by heating at 65°C for 10 min.

Recommended for:

  • Removal of mRNA prior to synthesis of second strand cDNA (1)
  • RT-PCR and qRT-PCR: removal of RNA after first strand cDNA synthesis
  • Removal of the poly(A) sequences of mRNA after hybridization with oligo(dT) (2)
  • Site-specific cleavage of RNA (3)
  • Studies of in vitro polyadenylation reaction products
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Enzyme RNase
Compatible Buffer 10X Reaction Buffer
Quantity 100 units
Product Type RNase H

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.