missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SdaI Buffer (10X)

Product Code. 10120880 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10120880 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10120880 Supplier Thermo Scientific™ Supplier No. B24

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Optimize digestions with SdaI restriction enzyme using this buffer, premixed with BSA for enhanced stability.

To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.

restriction enzymes exhibit 100% of their certified activity in the recommended buffer.

The following enzymes require unique buffers for optimal digestion: 

  • AarI 
  • AjiI 
  • BamHI 
  • BfuI 
  • Bpu10I 
  • BseXI 
  • Bsp143I 
  • Cfr9I 
  • Cfr10I 
  • Eam1105I 
  • Ecl136II 
  • Eco52I 
  • EcoRI 
  • KpnI 
  • Lsp1109I 
  • PacI 
  • PasI 
  • Ppu21I 
  • SacI 
  • ScaI 
  • SdaI 
  • SduI 
  • SgeI 
  • TaqI 
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Quantity 1 mL
Product Type SdaI Buffer
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.