Learn More
Descrizione
T4 RNA Ligase catalyzes the ATP-dependent intra- and intermolecular formation of phosphodiester bonds between 5'-phosphate and 3'-hydroxyl termini of oligonucleotides, single-stranded RNA and DNA.
The minimal substrate is a nucleoside 3',5'-biphosphate in intermolecular reaction and oligonucleotide of 8bases in intramolecular reaction.
- Source: E. coli cells with a cloned gene 63 of bacteriophage T4
- Molecular Weight: 43.6 kDa monomer
- The absence of ribonucleases, exodeoxyribonucleases, endodeoxyribonucleases and phosphatases confirmed by appropriate quality tests.
- E.coli cells with a cloned gene 63 of bacteriophage T4
Molecular Weight:
- 43.6kDa monomer
Definition of Activity Unit:
- One unit of the enzyme catalyzes the conversion of 1nmol of 5ft.-[32P]-(A)12-18 to a phosphatase-resistant form in 30 min. at 37°C
- Enzyme activity is assayed in the following mixture: 50mM Tris-HCl (pH 7.5), 10mM MgCl2, 10mM DTT, 1mM ATP, 10μM 5'-[32P]-(A)12-18 (10μM in 5ft.-termini)
Storage Buffer:
- The enzyme is supplied in:20mM Tris-HCl (pH 7.5), 1mM DTT, 50mM KCl, 0.1mM EDTA, 0.03% (v/v) ELUGENT Detergent and 50% (v/v) glycerol
10X Reaction Buffer:
- 500mM Tris-HCl (pH 7.5 at 25°C), 100mM MgCl2, 100mM DTT, 10 mM ATP
Inhibition and Inactivation:
- Inhibitors: metal chelators, SH group-modifying reagents (8).
- Inactivated by heating at 70°C for 10min.
Recommended for:
RNA 3'-end labeling with cytidine 3',5'-bis [alpha-32P] phosphate (1); Joining RNA to RNA (2); Synthesis of oligoribonucleotides and oligodeoxyribonucleotides (3, 4); Specific modifications of tRNAs (5); Oligodeoxyribonucleotide ligation to single-stranded cDNAs for 5ft. RACE (Rapid Amplification of cDNA Ends) (6); Site-specific generation of composite primers for PCR (7)
Note:
The recommended BSA concentration in the reaction mixture is 0.1mg/ml.
Specifica
Specifica
| Concentration | 10 U/μL |
| Enzyme | T4 RNA Ligase |
| Compatible Buffer | 10X Reaction Buffer |
| Quantity | 1,000 units |
| Product Type | T4 RNA Ligase |
For Research Use Only. Not for use in diagnostic procedures.
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.