missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Taq DNA Polymerase, recombinant (5 U/μL)

Product Code. 10349509 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 500 Units
100 Units
5 x 500 Units
500 Units
Unit Size:
100 units
500 units
Pack of 10
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10349509 5 x 500 Units Pack of 5
10374980 100 Units 100 units
10733941 500 Units 500 units
10272580 10 x 500 Units Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10349509 Supplier Thermo Scientific™ Supplier No. EP0405
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Thermo Scientific Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus.

Thermo Scientific Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5' to 3' synthesis of DNA, has no detectable 3' to 5' exonuclease (proofreading) activity, and possesses low 5' to 3' exonuclease activity. In addition, Taq DNA Polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Recombinant Taq DNA Polymerase is the ideal tool for standard PCR of templates 5 kb or shorter.

Features

  • Thermostable—half life is more than 40 min at 95°C
  • Generates PCR products with 3'-dA overhangs
  • Supplied with two buffers: 10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescent-labeled nucleotides)

Applications

  • Routine PCR amplification of DNA fragments up to 5 kb
  • High throughput PCR
  • DNA labeling

Notes

  • The error rate of Taq DNA Polymerase in PCR is 2.2 x 10-5 errors per nt per cycle. Accordingly, the accuracy of PCR is 4.5 x 104. Accuracy is an inverse of the error rate and shows an average number of correct nucleotides incorporated before an error occurs.
  • 10X Taq Buffer without Detergent is recommended for microarray experiments.
  • 10X Taq Buffer with (NH4)2SO4 allows for PCR at wide range of magnesium concentrations and decreases unspecific priming.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage Taq DNA Polymerase (5 U/μL), 5 x 100 μL
• 10X Taq Buffer with KCl, 10 x 1.25 mL
• 10X Taq Buffer with (NH4)2SO4, 10 x 1.25 mL
• 25 mM MgCl2, 10 x 1.25 mL

Store at -20°C.
GC-Rich PCR Performance Low
Polymerase Taq DNA Polymerase
Reaction Speed Standard
Product Type Taq DNA Polymerase (Recombinant)
Quantity 5 x 500 Units
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
Fidelity (vs. Taq) 1X
Hot Start No
Overhang 3'-A
Reaction Format Separate Components
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.