missing translation for 'onlineSavingsMsg'
Learn More

Ferrate(3-), tris[ethanedioato(2-)-κO1,κO2]-, potassium, hydrate (1:3:3), (OC-6-11)-, 97%, BLD Pharm™

Product Code. 31105244
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31105244 1 kg
31097427 100 g
31102303 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31105244 Supplier BLD Pharm Supplier No. BD1489421000g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5936-11-8
Molecular Formula C6H6FeK3O15
MDL Number MFCD00150450
Quantity 1 kg
SMILES O=C1[O-][Fe+3]23([O-]C(C([O-]2)=O)=O)([O-]C(C([O-]3)=O)=O)[O-]C1=O.[H]O[H].[H]O[H].[H]O[H].[K+].[K+].[K+]
Molecular Weight (g/mol) 491.24
Percent Purity 97%
Chemical Name or Material Ferrate(3-), tris[ethanedioato(2-)-κO1,κO2]-, potassium, hydrate (1:3:3), (OC-6-11)-
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.