missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Hexahydrate, ACS Reagent Grade, Ricca Chemical

Product Code. 31750880
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750880 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31750880 Supplier Ricca Chemical Company Supplier No. RDCF0200500B1
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 10025-77-1
Color Brown-Yellow
CAS Min % 98.0
CAS Max % 100.0
Physical Form Deliquescent Crystals
Quantity 500 g
Packaging White Poly Wide Mouth Bottle
Molecular Formula Cl3FeH12O6
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
IUPAC Name iron(3+) hexahydrate trichloride
Molecular Weight (g/mol) 270.29
Percent Purity 97 to 102%
Grade ACS
Chemical Name or Material Ferric Chloride, Hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.