missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Hexahydrate, (Crystalline),MP Biomedicals

Product Code. 11418801 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418801 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11418801 Supplier MP Biomedicals Supplier No. 0215350091
  • ONLINE EXCLUSIVE PRICE
    153.00 €
    144.90 € / 1kg
    Save 8.10 €
    5% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 10025-77-1
Physical Form Crystalline
Quantity 1 kg
Molecular Formula FeCl3 · 6H2O
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
IUPAC Name iron(3+) hexahydrate trichloride
Molecular Weight (g/mol) 270.29
Formula Weight 270.3
Chemical Name or Material Ferric Chloride Hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ferric Chloride Hexahydrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.