missing translation for 'onlineSavingsMsg'
Learn More

Ferric Nitrate Nonahydrate, >98%, (Crystalline/ACS Reagent Grade), MP Biomedicals

Product Code. 11440612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
11440612 500 g 500g
1 options
This item is not returnable. View return policy

Product Code. 11440612

Brand: MP Biomedicals 0219404690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7782-61-8
Physical Form Crystalline
Quantity 500 g
Assay Percent Range >98%
Molecular Formula FeH18N3O18
MDL Number MFCD00149708
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name iron(3+) nonahydrate trinitrate
Molecular Weight (g/mol) 403.99
PubChem CID 16211566
Formula Weight 404
Grade ACS Reagent
Chemical Name or Material Ferric Nitrate Nonahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.