missing translation for 'onlineSavingsMsg'
Learn More

Ferrocene, 99%

Product Code. 11364949
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
250 g
1 kg
missing translation for 'unitSize'
1kg
250g
50g
Product Code. Quantity unitSize
11364949 1 kg 1kg
11344949 50 g 50g
11354949 250 g 250g
3 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 11364949

missing translation for 'mfr': Thermo Scientific Alfa Aesar 087202.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102-54-5 | C10H10Fe | 186.04 g/mol

Ferrocene is used as a catalyst for carbon nanotubes and for rocket propellants. It is also used in the fuel for petrol engines, gasoline antiknock, luminous sensitive materials and exhibit anticancer activity. It acts as a photosensitive catalyst and promote the photochemical degadation of plastic films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-54-5
Molecular Formula C10H10Fe
Molecular Weight (g/mol) 186.04
MDL Number MFCD00001427
InChI Key DFRHTHSZMBROSH-UHFFFAOYSA-N
Synonym ferrocene, bis cyclopentadienyl iron
PubChem CID 25199998
IUPAC Name cyclopenta-1,3-diene;iron
SMILES [Fe].c1cccc1.c1cccc1

Specifications

Melting Point 173°C to 174°C
Color Yellow to Orange
Boiling Point 249°C
Odor Characteristic
Quantity 1 kg
UN Number UN1325
Sensitivity Air and moisture sensitive
Merck Index 14,4037
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 186.04
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ferrocene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.