missing translation for 'onlineSavingsMsg'
Learn More

Ferroin indicator solution, Solstice

Product Code. 15614240
Change view
Click to view available options
Quantity:
100 mL
500 mL
6 x 100 mL
6 x 500 mL
Unit Size:
100mL
500mL
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15614240 100 mL 100mL
15624240 500 mL 500mL
15634240 6 x 100 mL Pack of 6
15644240 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15614240 Supplier Honeywell-Fluka Supplier No. 34531100ML
  • ONLINE EXCLUSIVE PRICE
    71.00 €
    63.25 € / 100mL
    Save 7.75 €
    10% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Redox Indicator, Reag. Ph. Eur., E0 in sulfuric acid 1 mol/l = +1,06 volt

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14634-91-4
Molecular Formula C36H24FeN6O4S
Molecular Weight (g/mol) 692.53
MDL Number MFCD00036428
InChI Key CIWXFRVOSDNDJZ-UHFFFAOYSA-L
Synonym ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1
PubChem CID 84567
SMILES [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Quantity 100 mL
Linear Formula [Fe(C12H8N2)3]SO4
UN Number UN3082
IUPAC Name iron(2+);1,10-phenanthroline;sulfate
Formula Weight 692.52g/mol
Chemical Name or Material Ferroin indicator solution
Product Identifier
  • Ferroin indicator solution
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: 1,10-Phenanthrolinemonohydrochloride monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.