missing translation for 'onlineSavingsMsg'
Learn More

Ferroin Sulfate (0.025M aq solution) (Technical Grade), TRC

Product Code. 30393616
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393616 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393616 Supplier Toronto Research Chemicals Supplier No. TRCF30755050ML
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ferroin sulfate
CAS 14634-91-4
Molecular Formula C36 H24 Fe N6 . O4 S
Quantity 50 mL
Synonym Iron(2+), tris(1,10-phenanthroline-κN1,κN10)-, (OC-6-11)-, sulfate (1:1), Iron(2+), tris(1,10-phenanthroline)-, sulfate (1:1) (8CI), Iron(2+), tris(1,10-phenanthroline-N1,N10)-, (OC-6-11)-, sulfate (1:1), Tris(1,10-phenanthroline)iron sulfate (6CI,7CI), Ferroin sulfate, Tris(1,10-phenanthroline)iron(2+) sulfate (1:1), Tris(1,10-phenanthroline)iron(II) sulfate, Tris(o-phenanthroline)iron(2+) sulfate (1:1)
InChI Formula InChI=1S/3C12H8N2.Fe.H2O4S/c3*1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;1-5(2,3)4/h3*1-8H;;(H2,1,2,3,4)/q;;;+2;/p-2
SMILES [O-]S(=O)(=O)[O-].C1=Cc2ccc3C=CC=N4c3c2N(=C1)[Fe+2]456(N7=CC=Cc8ccc9C=CC=N5c9c78)N%10=CC=Cc%11ccc%12C=CC=N6c%12c%10%11
Molecular Weight (g/mol) 692.52
Formula Weight 692.0929
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.