missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate Hexahydrate, 98.5-101.5%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11422842 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422842 500 g 500g
11432842 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422842 Supplier MP Biomedicals Supplier No. 0215252390
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 7783-85-9
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
Assay Percent Range 98.5-101.5%
Molecular Formula Fe[NH4]2[SO4]2 · 6H2O
Formula Weight 392.1
Grade ACS Reagent
Chemical Name or Material Ferrous Ammonium Sulfate Hexahydrate
compliance-icons
Product Identifier
  • Ferrous Ammonium Sulfate Hexahydrate, ACS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ferrous Ammonium Sulfate Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.