missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate Heptahydrate, >99%, (Crystalline), MP Biomedicals

Product Code. 11415481 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415481 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11415481 Supplier MP Biomedicals Supplier No. 0215804883
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity crystalline powder intended for laboratory research applications.

Specifications

CAS 7782-63-0
Physical Form Crystalline
Quantity 250 g
Assay Percent Range >99%
Molecular Formula FeH14O11S
MDL Number MFCD00149719
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
IUPAC Name λ²-iron(2+) heptahydrate sulfate
Molecular Weight (g/mol) 278.01
PubChem CID 62662
ChEBI CHEBI:75836
Formula Weight 278
Percent Purity 99%
Chemical Name or Material Ferrous Sulfate Heptahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.