missing translation for 'onlineSavingsMsg'
Learn More

Ferrozine mono-sodium salt hydrate, 98%, BLD Pharm™

Product Code. 31100961
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31100961 25 g
31094493 5 g
31099702 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31100961 Supplier BLD Pharm Supplier No. BD0082776825g
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 63451-29-6
Molecular Formula C20H15N4NaO7S2
MDL Number MFCD00150794
Quantity 25 g
SMILES OS(=O)(C1=CC=C(C2=NN=C(C3=NC=CC=C3)N=C2C4=CC=C(S([O-])(=O)=O)C=C4)C=C1)=O.O.[Na+]
Molecular Weight (g/mol) 510.48
Percent Purity 98%
Chemical Name or Material Ferrozine mono-sodium salt hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.