missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals FerroZine™ Iron Reagent, Hydrate, 95+%, Pure

Product Code. 11976631 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11976631

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1266615-85-3 | C20H13N4NaO6S2 | 492.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-85-3
Molecular Formula C20H13N4NaO6S2
Molecular Weight (g/mol) 492.46
MDL Number MFCD00150794
InChI Key ZGVNYCXXBQPDPQ-UHFFFAOYSA-M
Synonym ferrozine mono-sodium salt hydrate, ferrozine™ iron reagent, c20h13n4o6s2.na.h2o, pdt disulfonate monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p inverted exclamation marka-disulfonic acid monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid monosodium salt hydrate
PubChem CID 129893581
IUPAC Name sodium 4-[3-(pyridin-2-yl)-6-(4-sulfophenyl)-1,2,4-triazin-5-yl]benzene-1-sulfonate
SMILES [Na+].OS(=O)(=O)C1=CC=C(C=C1)C1=C(N=C(N=N1)C1=CC=CC=N1)C1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >300.0°C
Color Yellow
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 95+%
Solubility Information Solubility in water: soluble
Formula Weight 492.46
Percent Purity ≥95%
Grade Pure
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material FerroZine* iron reagent,hydrate
Show More Show Less
compliance-icons
Product Identifier
  • FerroZine iron reagent, hydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.