missing translation for 'onlineSavingsMsg'
Learn More

Ferulic acid methyl ester, MedChemExpress

Product Code. 16489396
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16489396 Supplier Medchem Express Supplier No. HYW018643

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Ferulic acid methyl ester (Methyl ferulate) is a derivative of ferulic acid, isolated from Stemona tuberosa, with anti-inflammatory and antioxidant properties.

Ferulic acid methyl ester is a cell membrane and brain permeable compound, shows free radical scavenging ability, used in the research of neurodegenerative disorders. Ferulic acid methyl ester inhibits COX-2 expression, blocks p-p38 and p-JNK in primary bone marrow derived-macrophages.

Specifications

Chemical Name or Material Ferulic acid methyl ester
CAS 2309-07-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H12O4
Quantity 100 mg
Synonym Methyl ferulate
Solubility Information DMSO : ≥ 100 mg/mL (480.28 mM) ∣H2O : 2 mg/mL (9.61 mM; Need ultrasonic)
SMILES O=C(/C=C/C1=CC=C(C(OC)=C1)O)OC
Molecular Weight (g/mol) 208.21
Formula Weight 208.21
Percent Purity 99.18%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.