missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, charcoal stripped

Product Code. 15634559 Shop All Gibco Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
Format:
Bottle
One Shot
Unit Size:
500mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Unit Size
15634559 50 mL One Shot 50mL
10706143 500 mL Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15634559 Supplier Gibco™ Supplier No. A3382101
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays.

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Specialty
  • Serum Treatment: Stripped with charcoal-dextran
  • Recommended Storage: -5°C to -20°C

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Bovine
Age Fetal
Country of Origin Refer to CoA for specific origin
Form Liquid
Format One Shot
Product Type Fetal Bovine Serum
Quantity 50 mL
Serum Treatment Charcoal-stripped
Sterility Sterile
Product Line One Shot
Shipping Condition Dry Ice
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.