missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, Premium

Product Code. 17914671
Click to view available options
Quantity:
50 mL
500 mL
10 x 50 mL
Format:
Bottle
One Shot
Heat Inactivated:
No
Yes
Gamma-irradiated:
No
Yes
Unit Size:
500mL
50mL
Pack of 10
This item is not returnable. View return policy

Product Code. 17914671

Brand: Gibco™ A5670801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco fetal bovine sera offer an excellent value for basic cell culture, specialty research, and specific assays.

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Premium

  • Use for general cell culture with common cell lines
  • Low endotoxin and hemoglobin levels
  • Endotoxin level: <10 EU/mL
  • Hemoglobin level: <25 mg/dL
  • Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco serum delivers

  • ISO 13485–certified; processed in FDA registered facilities
  • Triple-filtered at 0.1 μm
  • Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

* FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at ≤-10°C.
Species Cattle/Bovine
Age Fetal
Country of Origin Refer to CoA for specific origin*
Form Frozen
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Heat-inactivated
Sterility Sterile-filtered
Sterilization Method Triple-filtered, 0.1 μm
Grade Premium
Tested For Up to 60 quality tests including 9CFR virus testing, endotoxin, and performance
Endotoxin Concentration ≤10 EU/mL
For Use With (Application) *FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.
Green Features Less waste
Heat Inactivated Yes
Hemoglobin Concentration ≤25 mg/dL
Purity or Quality Grade Premium
Shipping Condition Dry Ice
Gamma-irradiated No
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.