missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Fetal Bovine Serum, Premium, New Zealand Origin
BIOPHARMAGREENER_CHOICE

Product Code. 17162454
Change view
Click to view available options
Quantity:
500 mL
Serum Treatment:
Gamma-irradiated
Unit Size:
635g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Serum Treatment unitSize
17162454 500 mL Gamma-irradiated 635g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17162454 Supplier Corning™ Supplier No. 35098CV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Collected at government registered facilities in New Zealand

  • Exported from the country of origin accompanied with a Veterinarian Health Certificate and imported under a USDA-APHIS issued import permit.
  • True pooled and is triple 0.1 micron sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.
  • Validated process used to heat inactivate serum at 56°C ± 2°C for 30 minutes via bulk inactivation of entire lot in the pooling tank or individual bottles in a water bath.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -40°C to -10°C
Species Bovine
Country of Origin New Zealand
Form Liquid
Quantity 500 mL
Serum Treatment Gamma-irradiated
Sterility Sterile
Source New Zealand Origin
Heat Inactivated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.