missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, Premium Plus

Product Code. 30534604
Click to view available options
Quantity:
50 mL
500 mL
10 x 50 mL
Format:
Bottle
One Shot
Heat Inactivated:
No
Yes
Gamma-irradiated:
No
Yes
Unit Size:
500mL
50mL
Pack of 10
This item is not returnable. View return policy

Product Code. 30534604

Brand: Gibco™ A5669901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Premium Plus FBS, our most defined FBS, provides consistent cell growth for sensitive cell lines requiring the lowest viral risk.

Premium Plus FBS, our most defined FBS, provides consistent cell growth for sensitive cell lines requiring the lowest viral risk. It is ideal for use in biomanufacturing processes and diagnostic cell culture research requiring the most stringent quality profile, including an extensive biochemical and hormonal profile. Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Premium Plus

  • Use for a broad range of cell types, especially sensitive cell lines
  • Our most characterized serum with the lowest endotoxin and hemoglobin levels
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤20 mg/dL
  • Origin: Refer to CoA for specific origin (US, Australia, New Zealand)
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco serum delivers

  • ISO 13485-certified; processed in FDA-registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle easier to use in hood, reduces risk of contamination and helps improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

* FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.

Available in 30-45kGy; 25-40kGy Available upon request; 30-40kGy Available in the EU

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Species Cattle/Bovine
Age Fetal
Country of Origin Refer to CoA for specific origin*
Form Frozen
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Grade Premium Plus
Processing Triple filtered at 0.1 μm
Volume (Metric) 500 mL
Endotoxin Concentration ≤5 EU/mL
For Use With (Application) *FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.
Heat Inactivated No
Hemoglobin Concentration ≤20 mg/dL
Purity or Quality Grade Premium Plus
Product Line 26S
Shipping Condition Dry Ice
USDA Safety Testing Included USDA Safety Testing Performed for Australian origins.
Gamma-irradiated Yes
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.