missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, Brazil

Product Code. 11563397
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11563397

Brand: Gibco™ 10270098

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C results.

Sera Category: Standard
Origin: E.U. Approved (South American)
Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C results.
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Basic cell culture, specialty research, and specific assays
Certifications/Compliance EU-Approved
Content And Storage Store in freezer (–5 to –30°C)
Description Fetal Bovine Serum, E.U. Approved (South American Origin), 100mL, Liquid, –5 to –30°C Storage Temperature, Fetal, Cattle/Bovine Species, Standard (Sterile-Filtered) Serum Treatment, Dry Ice, Virus and Mycoplasma Tested
Country of Origin E.U. Approved (South American)
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 100 mL
Sterilization Method Sterile-filtered
Source Cattle/Bovine
Tested For Virus, Mycoplasma
Formulation Liquid
For Use With (Application) Basic cell culture, specialty research, and specific assays
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.