missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, heat inactivated, Australia

Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia orgin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

Brand:  Gibco™ 10100139

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11583387

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Sera Category: Secure
  • Origin: Australia
  • Endotoxin level: ≤10EU/mL
  • Hemoglobin level: ≤30mg/dL (levels routinely ≤25mg/dL)

Cell Culture, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Sf9 and Sf21 Cell Culture

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Liquid
Fetal Bovine Serum
Heat-inactivated
Sustainable packaging
Qualified
Frozen
Fetal
Australia
Bottle
100 mL
≤5 EU/mL
≤30 mg/dL
Gibco™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers