missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, heat inactivated, Brazil

Offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support

Brand:  Gibco™ A3840402

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15975161

  • 522.00 € / 10 units
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
  • Serum category: Value (Standard)
  • Serum for cell culture with robust cell lines
  • Good value for basic research
  • Endotoxin level: ≤ 50 EU/mL (levels routinely ≤ 10 EU/mL)
  • Hemoglobin level: ≤ 30 mg/dL
  • Origin: Brazil
  • ISO 13485-certified, processed in FDA-registered facilities
  • Labeled for in vitro diagnostics (compliant with highest level of USP sterility testing)
  • Triple filtered at 0.1 μm
  • Heat-inactivated serum in a One-Shot 50-mL bottle increases workflow benefits, while maintaining serum quality. The 50-mL bottle eliminates the need to aliquot and reduces the risk of contamination. Purchasing heat-inactivated serum eliminates the multiple steps necessary to heat inactivate and improves product integrity and consistency since the heat inactivation is done in a controlled environment.
  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label containing all of the information you need
  • 33% less waste generated than aliquoting from 500-mL bottles
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Bovine
One Shot
10 x 50 mL
Sterile
Triple Filtered at 0.1 μm
Qualified
Frozen
Fetal
Brazil
Fetal Bovine Serum
Heat-inactivated
Triple-filtered, 0.1 μm
10 x 50 mL
One Shot™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers