missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, heat inactivated, Brazil

Product Code. p-7181095
Change view
Click to view available options
Quantity:
50 mL
10 x 50 mL
Format:
One Shot
Unit Size:
1 bottle
10 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format unitSize
15961842 50 mL One Shot 1 bottle
15975161 10 x 50 mL One Shot 10 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15961842 Supplier Gibco™ Supplier No. A3840401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fetal Bovine Serum, qualified, heat inactivated, Brazil offers excellent value for basic cell culture, specialty research, and specific assays.

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Replacing with Fetal Bovine Serum, value
  • Sera application: for cell culture with robust cell lines, good value for basic research
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
  • Origin: Brazil

Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing QC results.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Bovine
Age Fetal
Country of Origin Brazil
Form Liquid
Format One Shot
Product Type Fetal Bovine Serum
Quantity 50 mL
Serum Treatment Heat-inactivated
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Purity or Quality Grade Qualified
Product Line One Shot
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.