missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, United States
GREENER_CHOICE

Product Code. 11560636
Click to view available options
Quantity:
50 mL
100 mL
500 mL
10 x 50 mL
Format:
Bottle
One Shot
Unit Size:
100mL
500mL
50mL
Pack of 10
This item is not returnable. View return policy

Product Code. 11560636

Brand: Gibco™ 26140079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Important Update: We've recently introduced Premium Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified US origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Gibco Sera: Trusted Quality for Consistent Performance

Gibco Sera Category: Value Plus (Performance)

  • Use for general cell culture with common cell lines
  • Low endotoxin and hemoglobin levels
  • Endotoxin level: <10 EU/mL
  • Hemoglobin level: <25 mg/dL
  • Origin: United States

Gibco Serum Delivers

  • ISO 13485 certified, processed in FDA registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age Fetal
Country of Origin United States
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Endotoxin Concentration ≤10 EU/mL
Hemoglobin Concentration ≤25 mg/dL
Purity or Quality Grade Qualified
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.