missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Fetal Bovine Serum (U.S.), Super Low IgG
Processed using protein G chromatography to reduce the levels to those required for monoclonal antibody production
175.00 € - 921.00 €
Product Code | Brand | Heat Inactivated | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Heat Inactivated | Quantity | Price | Quantity & Availability | ||||
16221122
|
Cytiva
SH30898.03IR25-40 |
No | 500 mL |
755.00 €
500mL |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11547310
|
Cytiva
SH30898.02 |
No | 100 mL |
175.00 €
100mL |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10329953
|
Cytiva
SH30898.03 |
No | 500 mL |
921.00 €
500mL |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12360143
|
Cytiva
SH30898.03IH |
Yes | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Fetal bovine serum (FBS) has natural levels of IgG that are too high for some cell culture applications. Lots can be prescreened to find the lowest naturally occuring levels, but as a biological product, FBS is frequently in short supply. Even lots with naturally low IgG levels may still contain too much for some applications. Examples of typical IgG levels in bovine serum products can be found in the table below.
- Performance is equivalent to non-processed FBS
- Suitable for applications where low levels of specific antibody are required (e.g. propagation of bovine viruses, monoclonal antibody production)
Specifications
2 to 8°C |