missing translation for 'onlineSavingsMsg'
Learn More

FG-2216, MedChemExpress

Product Code. p-200101368
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16482787

Brand: Medchem Express HY15641

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FG-2216 (IOX3) is a potent and orally active inhibitor of HIF prolyl hydroxylase-2 (PHD2), with an IC50 of 3.9 μM. FG-2216 induces robust erythropoietin and modest fetal hemoglobin in vivo.

FG-2216 (IOX3) is a potent and orally active inhibitor of HIF prolyl hydroxylase-2 (PHD2), with an IC50 of 3.9 μM. FG-2216 induces robust erythropoietin and modest fetal hemoglobin in vivo.

Specifications

Chemical Name or Material FG-2216
CAS 223387-75-5
Color White
Purity Grade Notes Research
Molecular Formula C12H9ClN2O4
Quantity 50 mg
Synonym IOX3 YM311
Solubility Information DMSO : ≥ 31 mg/mL (110.45 mM)
SMILES O=C(O)CNC(C1=C(O)C2=C(C(Cl)=N1)C=CC=C2)=O
Molecular Weight (g/mol) 280.66
Formula Weight 280.66
Percent Purity 99.32%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.