missing translation for 'onlineSavingsMsg'
Learn More

Fiacitabine, MedChemExpress

Product Code. 16460558
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460558 5 mg 5mg
16438098 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460558 Supplier Medchem Express Supplier No. HY50735
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fiacitabine(NSC 382097; FIAC; FOAC) is a selective inhibitior of DNA replication of herpes simplex virus(HSV) with IC50 values of 2.5 nM and 12.6 nM for HSV1 and HSV2, respectively.

Fiacitabine(NSC 382097

Specifications

Chemical Name or Material Fiacitabine
CAS 69123-90-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C9H11FIN3O4
Quantity 5 mg
Synonym NSC 382097 FIAC FOAC
Solubility Information DMSO : ≥ 37 mg/mL (99.70 mM)
SMILES O=C(N=C1N)N([C@@H]([C@H]([C@@H]2O)F)O[C@@H]2CO)C=C1I
Molecular Weight (g/mol) 371.1
Formula Weight 371.1
Percent Purity 98.83%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.