Learn More
Thermo Scientific™ Fiberlite F12-6x500 LEX Rotor O-ring Refresher Kit with Grease
Product Code. 15772292
Change view
Click to view available options
For Use With (Equipment):
F12-6x500 LEX Rotor
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15772292 F12-6x500 LEX Rotor Each
1 options
Product Code. 15772292 Supplier Thermo Scientific™ Supplier No. 021062375
  • ONLINE EXCLUSIVE PRICE
    175.88 €
    142.13 € / Pack of 1
    Save 33.75 €
    19% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes five Replacement O-rings and and one O-Ring Grease Packet for rotor sealing system.
Includes five Replacement O-rings and and one O-Ring Grease Packet for rotor sealing system.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) F12-6x500 LEX Rotor
Quantity 5 Pk.
Product Type O-Ring