missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fiberlite F6-10 x 1000 LEX Carbon Fiber Rotor (for BIOS A Large-Capacity Centrifuge)

Product Code. 15995350 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
10 x 1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15995350 10 x 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15995350 Supplier Thermo Scientific™ Supplier No. 096101001
  • ONLINE EXCLUSIVE PRICE
    43590.00 €
    36270.00 € / Pack of 1
    Save 7320.00 €
    16% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accelerate bioprocessing with the 10-liter capacity and an RCF of up to 12,000 x G of the Thermo Scientific™ Fiberlite™ F6-10x1000 LEX rotor.

Batch biprocessing with the Thermo Scientific™ Sorvall™ RC BIOS A centrifuge system.

High throughput reduces cycle times by spinning up to 10 x 1 liter bottles in a single spin.

Enhance productivity with Fiberlite carbon fiber rotors

  • Unequalled structural integrity:
    • Fatigue resistance eliminates the need to derate or limit the speed over the rotor lifespan
    • Corrosion-resistant design protects against ever-present hazard due to moisture, chemicals, alkaline solutions or other salts in the laboratory
  • Load tubes directly into the rotor, eliminating multi-canister assemblies, which can be misplaced or damaged
  • Secure investment, backed by a 15-year warranty

Ordering information: Order rotor body (096-01001) for complete assembly. Rotor Body comes with a set of Thermo Scientific Fiberlite Superspeed PPCO bottles 010-1491. For rotor care kit, order 021-101001; Lid assembly, order 099-101001;  Contact your sales representative for information.

Compatible with: Thermo Scientific™ Sorvall™ RC BIOS A, Thermo Scientific™ Fiberlite™ superspeed bottles (010-1491 and 0101-1492)

TRUSTED_SUSTAINABILITY

Specifications

Description Fiberlite F6-10x1000 LEX Carbon Fiber Rotor
Material Carbon Fiber
Type Rotor
Capacity 10 x 1000 mL
Max. RCF 12,000 x g
Max. Speed 6,250 rpm
Radius (Metric) 274 mm
Radius (Metric) Min. 124 mm
K Factor 5135
Max. Tube Length 195 mm
Max. Tube Diameter 98 mm
Net Weight 31.7 lb.
For Use With (Equipment) Sorvall BIOS A Centrifuge Heavy Duty
Angle 25°
Avg. RCF 12,000 x g
Includes Complete with set of 10 bottles
No. of Places 10
Biocontainment No
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.