missing translation for 'onlineSavingsMsg'
Learn More

Fiboflapon, MedChemExpress

Product Code. 16462947
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16462947

Brand: Medchem Express HY15874

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fiboflapon (GSK2190915; AM-803) is a potent and orally bioavailable 5-lipoxygenase-activating protein (FLAP) inhibitor with a potency of 2.9 nM in FLAP binding, an IC50 of 76 nM for inhibition of LTB4 in human blood.

Fiboflapon (GSK2190915

Specifications

Chemical Name or Material Fiboflapon
CAS 936350-00-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C38H43N3O4S
Quantity 50 mg
Synonym GSK2190915 AM-803
Solubility Information DMSO : 50 mg/mL (78.39 mM; Need ultrasonic)
SMILES CC(C(O)=O)(C)CC1=C(SC(C)(C)C)C2=CC(OCC3=CC=C(C)C=N3)=CC=C2N1CC4=CC=C(C5=CN=C(OCC)C=C5)C=C4
Molecular Weight (g/mol) 637.83
Formula Weight 637.83
Percent Purity 98.54%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.