missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fibroblast Growth Factor, basic, mouse

Product Code. 15438689
Change view
Click to view available options
Quantity:
10μg
50μg
Unit Size:
10µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438689 50μg 50µg
15428689 10μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438689 Supplier Thermo Scientific Chemicals Supplier No. J67243.I&
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 16.2 kd
Quantity 50μg
Packaging Plastic bottle
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.