missing translation for 'onlineSavingsMsg'
Learn More

Ficodox™ Plus, Pure, Mixed COD Reagent, Fisher Chemical™

Product Code. 10437450 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10437450

Brand: Fisher Chemical J/4010/15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

MDL Number 283231

Specifications

Physical Form Liquid
Melting Point 3°C
Boiling Point 280°C
Packaging Plastic Coated Glass bottle
Color Brown to Yellow
Quantity 1 L
compliance-icons compliance-icons
Product Identifier
  • Ficodox]TS Plus
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.